Novus Biologicals
Manufacturer Code:NBP243566AF594
Catalog # NP24366A94
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3611 |
Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of human ARID1A. The exact sequence is proprietary. |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: ARID domain-containing protein 1A; ARID domain-containing protein 1A AT rich interactive domain 1A (SWI- like) AT rich interactive domain 1A (SWI-like) AT-rich interactive domain-containing protein 1A B120SWI-like protein BAF250a BAF250SWI/SNF complex protein p270 BM029 brain protein 120 BRG1-associated factor 250 BRG1-associated factor 250a C10rf4 C1orf4SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily F member 1 chromatin remodeling factor p250 hELD hOSA1 matrix associated actin dependent regulator of chromatin Osa homolog 1 OSA1 OSA1 nuclear protein P270 SMARCF1 subfamily f member 1; AT rich interactive domain 1A (SWI-like); AT-rich interactive domain-containing protein 1A; B120; BAF250; BAF250A; brain protein 120; BRG1-associated factor 250; BRG1-associated factor 250a; chromatin remodeling factor p250; hELD; hOSA1; Osa homolog 1; OSA1 nuclear protein; SWI-like protein; SWI/SNF complex protein p270; SWI/SNF-related, matrix-associated, actin-dependent regulator of chromatin subfamily F member 1
Gene Aliases: ARID1A; B120; BAF250; BAF250A; BM029; C1orf4; CSS2; ELD; hELD; hOSA1; MRD14; OSA1; P270; SMARCF1
UniProt ID: (Human) O14497
Entrez Gene ID: (Human) 8289
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.