Novus Biologicals
Manufacturer Code:NBP262217AF594
Catalog # NB002654
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | ARHGEF-08 |
Immunogen | Recombinant fragment of human ARHGEF4 (amino acids 143-271) |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: APC-stimulated guanine nucleotide exchange factor 1; APC-stimulated guanine nucleotide exchange factor 1 Asef ASEF1 ASEFAPC-stimulated guanine nucleotide exchange factor GEF4 KIAA1112 Rho guanine nucleotide exchange factor (GEF) 4 rho guanine nucleotide exchange factor 4 STM6; Asef; Rho guanine nucleotide exchange factor (GEF) 4; Rho guanine nucleotide exchange factor 4
Gene Aliases: ARHGEF4; ASEF; ASEF1; GEF4; KIAA1112; STM6
UniProt ID: (Human) Q9NR80
Entrez Gene ID: (Human) 50649
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.