Invitrogen
Manufacturer Code:PA570425
Catalog # PIPA570425
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of mouse ARHGEF4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: APC-stimulated guanine nucleotide exchange factor; APC-stimulated guanine nucleotide exchange factor 1; ARHGEF4 Rho guanine nucleotide exchange factor (GEF) 4 ASEF GEF4 STM6 KIAA1112 DKFZp434G2016 APC-stimulated guanine nucleotide exchange factor ASEF1 Rho guanine nucleotide exchange factor 4; Asef; Rho guanine nucleotide exchange factor (GEF) 4; Rho guanine nucleotide exchange factor 4
Gene Aliases: 9330140K16Rik; ARHGEF4; ASEF; ASEF1; ENSMUSG00000070955; GEF4; KIAA1112; STM6
UniProt ID: (Human) Q9NR80, (Mouse) Q7TNR9
Entrez Gene ID: (Rabbit) 100338395, (Dog) 483856, (Bovine) 505852, (Human) 50649, (Rat) 301334, (Mouse) 226970, (Guinea pig) 100728096, (Horse) 100067370
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.