Proteintech
Manufacturer Code:552131AP150UL
Catalog # 50-173-6060
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: APC-stimulated guanine nucleotide exchange factor; APC-stimulated guanine nucleotide exchange factor 1; ARHGEF4 ASEF ASEF1 GEF4 KIAA1112 STM6; Asef; Rho guanine nucleotide exchange factor (GEF) 4; Rho guanine nucleotide exchange factor 4
Gene Aliases: 9330140K16Rik; ARHGEF4; ASEF; ASEF1; ENSMUSG00000070955; GEF4; KIAA1112; STM6
UniProt ID: (Human) Q9NR80, (Mouse) Q7TNR9
Entrez Gene ID: (Human) 50649, (Mouse) 226970
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.