OriGene
Manufacturer Code:CF809320
Catalog # 50-166-9410
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5F2 |
Immunogen | Full length human recombit protein of human ARHGEF2 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: GEF GEF-H1 GEFH1 LFP40 NEDMHM P40; GEF-H1; Guanine nucleotide exchange factor H1; Microtubule-regulated Rho-GEF; Proliferating cell nucleolar antigen p40; Rho guanine nucleotide exchange factor 2; Rho/Rac guanine nucleotide exchange factor (GEF) 2
Gene Aliases: ARHGEF2; GEF; GEF-H1; GEFH1; KIAA0651; LFP40; P40
UniProt ID: (Human) Q8WUG4
Entrez Gene ID: (Human) 9181
Molecular Function:
G-protein modulator
enzyme modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.