Proteintech
Manufacturer Code:142821AP150UL
Catalog # 50-172-9340
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ARHGDIG Fusion Protein Ag5554 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARHGDIG Rho GDI 3 Rho GDI gamma RHOGDI 3; guanosine diphosphate (GDP) dissociation inhibitor 5, Rho associated; Rho GDI 3; Rho GDP dissociation inhibitor (GDI) gamma; Rho GDP-dissociation inhibitor 3; Rho-GDI gamma; Rho-GDI2; RhoGDI gamma
Gene Aliases: ARHGDIG; Gdi5; Rho-GDI-3; Rho-GDI2; RHOGDI-3; RIP2
UniProt ID: (Human) Q99819, (Mouse) Q62160
Entrez Gene ID: (Human) 398, (Rat) 360500, (Mouse) 14570
Molecular Function:
G-protein modulator
enzyme modulator
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.