Abnova
Manufacturer Code:H00000397A01
Catalog # 89-104-108
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 1 a.a. to 201 a.a) full-length recombinant protein with GST tag.,ARHGDIB (AAH09200 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length recombinant ARHGDIB.
For Research Use Only
Protein Aliases: D4; GDIA2; GDID4; Ly-GDI; LYGDI; RAP1GN1; Rho GDI 2; Rho GDP dissociation inhibitor (GDI) beta; Rho GDP-dissociation inhibitor 2; Rho-GDI beta; RhoGDI2
Gene Aliases: ARHGDIB; D4; GDIA2; GDID4; Ly-GDI; LYGDI; RAP1GN1; RhoGDI2
UniProt ID: (Human) P52566
Entrez Gene ID: (Human) 397
Molecular Function:
G-protein modulator
enzyme modulator
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.