OriGene
Manufacturer Code:CF501089
Catalog # 50-166-0127
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1F2 |
Immunogen | Full length human recombit protein of human ARHGDIA produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: arhgdia {ECO:0000312|RGD:1359547}; epididymis secretory sperm binding protein Li 47e; GDIA1 HEL-S-47e NPHS8 RHOGDI RHOGDI-1; GDP-dissociation inhibitor, aplysia RAS-related 1; Rho GDI 1; rho GDI 1 {ECO:0000250|UniProtKB:P19803}; Rho GDP dissociation inhibitor (GDI) alpha; Rho GDP-dissociation inhibitor 1; rho GDP-dissociation inhibitor 1 {ECO:0000250|UniProtKB:P19803}; Rho-GDI alpha; rho-GDI alpha {ECO:0000250|UniProtKB:P19803}
Gene Aliases: ARHGDIA; Bles01; GDIA1; HEL-S-47e; NPHS8; RHOGDI; RHOGDI-1
UniProt ID: (Human) P52565, (Rat) Q5XI73
Entrez Gene ID: (Human) 396, (Dog) 475924, (Rat) 360678
Molecular Function: G-protein modulator enzyme modulator signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.