Proteintech
Manufacturer Code:188341AP150UL
Catalog # 50-173-2250
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARHGAP24 Filamin A associated RhoGAP FILGAP p73 p73RhoGAP RC GAP72 RCGAP72 Rho type GTPase activating protein 24 RhoGAP of 73 kDa Sarcoma antigen NY SAR 88; Down-regulated in nephrectomized rat kidney #2; Filamin-A-associated RhoGAP; FilGAP; p73RhoGAP; RAC1- and CDC42-specific GTPase-activating protein of 72 kDa; RC-GAP72; Rho GTPase-activating protein 24; Rho-type GTPase-activating protein 24; RhoGAP of 73 kDa; Sarcoma antigen NY-SAR-88
Gene Aliases: 0610025G21Rik; ARHGAP24; DR-NR#2; FILGAP; p73; p73RhoGAP; RC-GAP72; RCGAP72; si:ch211-65i12.2
UniProt ID: (Human) Q8N264, (Mouse) Q8C4V1, (Rat) Q5U2Z7
Entrez Gene ID: (Human) 83478, (Dog) 478463, (Mouse) 231532, (Rat) 305156, (Zebrafish) 100005116
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.