Novus Biologicals
Manufacturer Code:NBP245615
Catalog # NBP245615
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7A5 |
Immunogen | Full length human recombinant protein of human LDLRAP1(NP_056442) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ARH1 ARH2 ARHAutosomal recessive hypercholesterolemia protein DKFZp586D0624 FHCB1ARH1 FHCB2LDL receptor adaptor protein low density lipoprotein receptor adapter protein 1 low density lipoprotein receptor adaptor protein 1 MGC34705; Autosomal recessive hypercholesterolemia protein; LDL receptor adaptor protein; Low density lipoprotein receptor adapter protein 1
Gene Aliases: ARH; ARH1; ARH2; FHCB1; FHCB2; LDLRAP1
UniProt ID: (Human) Q5SW96
Entrez Gene ID: (Human) 26119
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.