Proteintech
Manufacturer Code:132131AP150UL
Catalog # 50-172-8577
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ARH Fusion Protein Ag4006 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARH ARH1 ARH2 FHCB1 FHCB2 LDLRAP1; Autosomal recessive hypercholesterolemia protein; Autosomal recessive hypercholesterolemia protein homolog; LDL receptor adaptor protein; Low density lipoprotein receptor adapter protein 1
Gene Aliases: AA691260; ARH; ARH1; ARH2; FHCB1; FHCB2; LDLRAP1; RGD1563417
UniProt ID: (Human) Q5SW96, (Mouse) Q8C142
Entrez Gene ID: (Human) 26119, (Rat) 500564, (Mouse) 100017
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.