Invitrogen
Manufacturer Code:PA5102967
Catalog # PIPA5102967
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ARFRP1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1500006I01Rik ADP ribosylation factor related protein 1 ADP-ribosylation factor related protein 1 ADP-ribosylation factor-related protein 1 AI480700 ARF-related protein 1 Arfrp1 ARL18 ARP Arp1 helicase-like protein NHL SCG10 like-protein; ADP-ribosylation factor related protein 1; ADP-ribosylation factor-related protein 1; ARF-related protein 1; helicase-like protein NHL; SCG10 like-protein
Gene Aliases: ARFRP1; ARL18; ARP; ARP1
UniProt ID: (Human) Q13795
Entrez Gene ID: (Human) 10139
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.