Novus Biologicals
Manufacturer Code:NBP214306
Catalog # NBP214306
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: ANKQDVETCLSIPDIKTAFSDCTSKIGRRDCLTQACSALTGKGVREGIEW MVKCVVRNVHRPPRQRDIT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosylation factor related protein 1; ADP-ribosylation factor related protein 1 ARF-related protein 1 ARL18 ARP1 ARPADP-ribosylation factor-related protein 1 helicase-like protein NHL SCG10 like-protein; ADP-ribosylation factor-related protein 1; ARF-related protein 1; helicase-like protein NHL; SCG10 like-protein
Gene Aliases: ARFRP1; ARL18; ARP; ARP1
UniProt ID: (Human) Q13795
Entrez Gene ID: (Human) 10139
Molecular Function:
G-protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.