Proteintech
Manufacturer Code:177121AP150UL
Catalog # 50-173-1735
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ARFRP1 Fusion Protein Ag12036 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosylation factor related protein 1; ADP-ribosylation factor-related protein 1; ARF related protein 1 ARFRP1 ARL18 ARP Arp1; ARF-related protein 1; ARP; helicase-like protein NHL; SCG10 like-protein
Gene Aliases: 1500006I01Rik; AI480700; ARFRP1; ARL18; ARP; ARP1
UniProt ID: (Human) Q13795, (Mouse) Q8BXL7, (Rat) Q63055
Entrez Gene ID: (Human) 10139, (Mouse) 76688, (Rat) 117051
Molecular Function:
G-protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.