Invitrogen
Manufacturer Code:PA545214
Catalog # PIPA545214
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human ARFGEF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosylation factor guanine nucleotide-exchange factor 1; ADP-ribosylation factor guanine nucleotide-exchange factor 1 (brefeldin A-inhibited); ADP-ribosylation factor guanine nucleotide-exchange factor 1(brefeldin A-inhibited); BIG1; Brefeldin A-inhibited GEP 1; Brefeldin A-inhibited guanine nucleotide-exchange protein 1; p200 ARF guanine nucleotide exchange factor; p200 ARF-GEP1
Gene Aliases: ARFGEF1; ARFGEP1; BIG1; D130059B05Rik; D730028O18Rik; hm:zeh1651; im:7138204; P200; RGD1560793; wu:fj64h08; zeh1651
UniProt ID: (Human) Q9Y6D6, (Bovine) O46382, (Rat) D4A631, (Mouse) G3X9K3
Entrez Gene ID: (Human) 10565, (Bovine) 281022, (Dog) 477903, (Horse) 100063272, (Rabbit) 100345151, (Rat) 312915, (Mouse) 211673, (Guinea pig) 100714174, (Zebrafish) 100329953
Molecular Function:
G-protein modulator
enzyme modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.