Novus Biologicals
Manufacturer Code:NBP200513
Catalog # NBP200513
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1F6 |
Immunogen | Full length human recombinant protein of human ARFGAP1 (NP_783202) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosylation factor 1 GTPase activating protein ADP-ribosylation factor 1 GTPase-activating protein ADP-ribosylation factor GTPase activating protein 1 ADP-ribosylation factor GTPase-activating protein 1 ARF GAP 1 ARF1 GAP ARF1-directed GTPase-activating protein ARF1GAP bA261N11.3 FLJ10767 GAP protein HRIHFB2281 MGC39924; ADP-ribosylation factor 1 GTPase-activating protein; ADP-ribosylation factor GTPase activating protein 1; ADP-ribosylation factor GTPase-activating protein 1; ARF GAP 1; ARF1 GAP; ARF1-directed GTPase-activating protein
Gene Aliases: ARF1GAP; ARFGAP1; HRIHFB2281
UniProt ID: (Human) Q8N6T3
Entrez Gene ID: (Human) 55738
Molecular Function: G-protein modulator enzyme modulator nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.