Invitrogen
Manufacturer Code:PA544191
Catalog # PIPA544191
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human ARF6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosylation factor 6; ADP-ribosylation factor 6 DKFZp564M0264
Gene Aliases: AI788669; ARF6; AW496366
UniProt ID: (Human) P62330, (Rat) P62332, (Mouse) P62331
Entrez Gene ID: (Sheep) 100302306, (Human) 382, (Horse) 100065920, (Dog) 490679, (Rabbit) 100345872, (Rat) 79121, (Mouse) 11845, (Guinea pig) 100726124
Molecular Function:
G-protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.