Invitrogen
Manufacturer Code:PA518627
Catalog # PIPA518627
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (EGLDWLSNQLRNQK) corresponding to the C-terminus amino acids of ARF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosylation factor 1; ADP-ribosylation factor 2; ADP-ribosylation factor 3; ADP-ribosylation factor 4; ARF1 ARF2 ARF3 ARF4 ARF1 ADP-ribosylation factor 1 ADP-ribosylation factor 2 ADP-ribosylation factor 3 ADP-ribosylation factor 4 pan ARF; Liver regeneration-related protein LRRG202; small GTP binding protein
Gene Aliases: 5430400P17Rik; AA407803; Ac1-253; AI854770; ARF1; ARF2; ARF3; ARF4
UniProt ID: (Human) P84077, (Human) P61204, (Human) P18085, (Mouse) P84078, (Rat) P84079, (Mouse) P10947, (Rat) P61206, (Mouse) P61205, (Rat) P61751, (Mouse) P61750
Entrez Gene ID: (Human) 375, (Human) 377, (Human) 378, (Mouse) 11840, (Rat) 64310, (Mouse) 11841, (Rat) 140940, (Mouse) 11842, (Rat) 79120, (Mouse) 11843
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.