Abnova
Manufacturer Code:H00008260A01
Catalog # 89-105-825
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 1 a.a. to 235 a.a) full-length recombinant protein with GST tag.,ARD1A (AAH00308 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length recombinant ARD1A.
For Research Use Only
Protein Aliases: ARD1; ARD1 homolog A, N-acetyltransferase; DXS707; hARD1; MGC71248; N-acetyltransferase ARD1, human homolog of; N-alpha-acetyltransferase 10; N-alpha-acetyltransferase 10, NatA catalytic subunit; N-terminal acetyltransferase complex ARD1 subunit homolog A; NatA catalytic subunit Naa10; TE2
Gene Aliases: ARD1; ARD1A; ARD1P; DXS707; MCOPS1; NAA10; NATD; OGDNS; TE2
UniProt ID: (Human) P41227
Entrez Gene ID: (Human) 8260
Molecular Function: acetyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.