Proteintech
Manufacturer Code:148031AP150UL
Catalog # 50-172-9734
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ARD1A Fusion Protein Ag6519 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha-N-acetyltransferase 1A; ARD1 ARD1A DXS707 NAA10 TE2; ARD1 homolog A, N-acetyltransferase; hARD1; N(alpha)-acetyltransferase 10, NatA catalytic subunitNalpha acetyltransferase 10; N-acetyltransferase ARD1 homolog; N-acetyltransferase ARD1, human homolog of; N-alpha-acetyltransferase 10; N-alpha-acetyltransferase 10, NatA catalytic subunit; N-terminal acetyltransferase complex ARD1 subunit homolog A; Nalpha acetyltransferase 10; NatA catalytic subunit Naa10
Gene Aliases: 2310039H09Rik; ARD1; ARD1A; ARD1P; DXS707; MCOPS1; NAA10; NATD; OGDNS; RGD1565315; TE2
UniProt ID: (Human) P41227, (Mouse) Q9QY36
Entrez Gene ID: (Human) 8260, (Rat) 363518, (Mouse) 56292
Molecular Function:
acetyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.