OriGene
Manufacturer Code:CF504314
Catalog # 50-166-2800
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4H9 |
Immunogen | Full length human recombit protein of human ARCN1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Archain; archain vesicle transport protein 1; coatomer delta subunit; coatomer protein complex, subunit delta; coatomer protein delta-COP; Coatomer subunit delta; COPD SRMMD; Delta-coat protein; Delta-COP
Gene Aliases: ARCN1; COPD
UniProt ID: (Human) P48444, (Rat) Q66H80
Entrez Gene ID: (Dog) 479415, (Human) 372, (Rat) 300674
Molecular Function:
membrane traffic protein
vesicle coat protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.