Novus Biologicals
Manufacturer Code:NB500257
Catalog # NB500257
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to residues 125-141 (PELQTLTRPGALRAPRR) of the human Nop30 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Apoptosis repressor with CARD; Apoptosis repressor with CARD ARCFLJ35304 CARD2 Muscle-enriched cytoplasmic protein MYC MYP NOP30 NOPnucleolar protein 3 nucleolar protein 3 (apoptosis repressor with CARD domain) Nucleolar protein of 30 kDa; Muscle-enriched cytoplasmic protein; Myp; Nop30; Nucleolar protein 3; nucleolar protein 3 (apoptosis repressor with CARD domain); Nucleolar protein of 30 kDa
Gene Aliases: ARC; FCM; MYP; NOL3; NOP; NOP30
UniProt ID: (Human) O60936
Entrez Gene ID: (Human) 8996
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.