Invitrogen
Manufacturer Code:PA520305
Catalog # PIPA520305
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 14 amino acid peptide from near the carboxy terminus of human Nop30. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Apoptosis repressor with CARD; ARC MYP NOP NOP30 muscle-enriched cytoplasmic protein nucleolar protein 3 nucleolar protein of 30 kDa; Muscle-enriched cytoplasmic protein; Myp; Nop30; Nucleolar protein 3; nucleolar protein 3 (apoptosis repressor with CARD domain); Nucleolar protein of 30 kDa
Gene Aliases: ARC; B430311C09Rik; FCM; MYC; MYP; NOL3; NOP; NOP30
UniProt ID: (Human) O60936, (Rat) Q62881, (Mouse) Q9D1X0
Entrez Gene ID: (Human) 8996, (Rat) 85383, (Mouse) 78688
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.