Invitrogen
Manufacturer Code:PA560158
Catalog # PIPA560158
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ARAP3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Arf and Rho GAP adapter protein 3; Arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 3; ARF-GAP, RHO-GAP, ankyrin repeat and plekstrin homology domains-containing protein 3; centaurin, delta 3; Centaurin-delta-3; CENTD3 centaurin delta 3 ARAP3 FLJ21065 ARF-GAP RHO-GAP ankyrin repeat and plekstrin homology domains-containing protein 3 PtdIns(345)P3-binding protein phosphoinositide binding protein ArfGAP with RhoGAP domain ankyrin repeat and PH domain 3 DRAG1 Arf and Rho GAP adapter protein 3; Cnt-d3; phosphoinositide binding protein; PtdIns(3,4,5)P3-binding protein
Gene Aliases: ARAP3; CENTD3; DRAG1
UniProt ID: (Human) Q8WWN8
Entrez Gene ID: (Human) 64411
Molecular Function: G-protein modulator enzyme modulator nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.