OriGene
Manufacturer Code:CF809124
Catalog # 50-166-9287
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3C5 |
Immunogen | Human recombit protein fragment corresponding to amino acids 420-659 of human AR produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AIS AR8 DHTR HUMARA HYSP1 KD NR3C4 SBMA SMAX1 TFM; androgen nuclear receptor variant 2; Androgen receptor; androgen receptor splice variant 4b; Dihydrotestosterone receptor; Nuclear receptor subfamily 3 group C member 4
Gene Aliases: AIS; AR; AR8; DHTR; HUMARA; HYSP1; KD; NR3C4; SBMA; SMAX1; TFM
UniProt ID: (Human) P10275
Entrez Gene ID: (Human) 367
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.