Abnova
Manufacturer Code:MAB9567
Catalog # 89-113-691
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AR27 |
Immunogen | Recombinant protein corresponding to 321 amino acids near N-terminus of human AR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C on dry atmosphere. After reconstitution with sterile distilled water,store at -20°C or below. Aliquot to avoid repeated freezing and thawing.,store at 4°C for at least two months. For long term storage |
Mouse monoclonal antibody raised against recombinant AR.
For Research Use Only
Protein Aliases: AIS; androgen nuclear receptor variant 2; Androgen receptor; androgen receptor splice variant 4b; DHTR; Dihydrotestosterone receptor; HUMARA; KD; NR3C4; Nuclear receptor subfamily 3 group C member 4; SBMA; SMAX1; TFM
Gene Aliases: AIS; AR; AR8; DHTR; HUMARA; HYSP1; KD; NR3C4; SBMA; SMAX1; TFM
UniProt ID: (Human) P10275
Entrez Gene ID: (Human) 367
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.