Proteintech
Manufacturer Code:251311AP150UL
Catalog # 50-173-4739
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | AQP7 Fusion Protein Ag17945 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AQP 7 AQP7 AQP7L AQP9 AQPap Aquaglyceroporin 7 aquaporin 7 Aquaporin 7 like Aquaporin adipose; AQP-7; AQPap; Aquaglyceroporin-7; Aquaporin adipose; Aquaporin-7; Aquaporin-7-like
Gene Aliases: AQP7; AQP7L; AQP9; AQPap; GLYCQTL
UniProt ID: (Human) O14520, (Mouse) O54794
Entrez Gene ID: (Human) 364, (Mouse) 11832
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.