Abnova
Manufacturer Code:MAB13009
Catalog # 89-933-579
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1F3 |
Immunogen | Recombinant protein corresponding to full length human AQP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against full length recombinant human AQP1.
For Research Use Only
Protein Aliases: AQP-1; AQP-CHIP; aquaporin 1 (channel-forming integral protein, 28kDa, CO blood group); aquaporin 1, Colton blood group antigen; Aquaporin-1; Aquaporin-CHIP; channel-like integral membrane protein, 28-kDa; CHIP28; CO; MGC26324; Urine water channel; Water channel protein for red blood cells and kidney proximal tubule
Gene Aliases: AQP-CHIP; AQP1; CHIP28; CO
UniProt ID: (Human) P29972
Entrez Gene ID: (Human) 358
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.