Invitrogen
Manufacturer Code:PA5102774
Catalog # PIPA5102774
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human APRT corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenine phosphoribosyl transferase adenine phosphoribosyltransferase AMP AMP diphosphorylase AMP pyrophosphorylase Aprt APRTD C85684 DKFZp686D13177 MGC125856 MGC125857 MGC129961 transphosphoribosidase; Adenine phosphoribosyltransferase; AMP diphosphorylase; AMP pyrophosphorylase; APRT; transphosphoribosidase
Gene Aliases: AMP; APRT; APRTD; C85684
UniProt ID: (Human) P07741, (Mouse) P08030, (Rat) P36972
Entrez Gene ID: (Human) 353, (Mouse) 11821, (Rat) 292072
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.