Novus Biologicals
Manufacturer Code:NBP15530020UL
Catalog # NBP15530020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to APRT(adenine phosphoribosyltransferase) The peptide sequence was selected from the N terminal of APRT. Peptide sequence ADSELQLVEQRIRSFPDFPTPGVVFRDISPVLKDPASFRAAIGLLARHLK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenine phosphoribosyltransferase; adenine phosphoribosyltransferase AMP AMP diphosphorylase AMP pyrophosphorylase DKFZp686D13177 EC 2.4.2.7 MGC125856 MGC125857 MGC129961 transphosphoribosidase; AMP diphosphorylase; AMP pyrophosphorylase; APRT; transphosphoribosidase
Gene Aliases: AMP; APRT; APRTD
UniProt ID: (Human) P07741
Entrez Gene ID: (Human) 353
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.