Novus Biologicals
Manufacturer Code:NBP292654100UL
Catalog # NB027177
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human APRT (NP_000476.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenine phosphoribosyltransferase; adenine phosphoribosyltransferase AMP AMP diphosphorylase AMP pyrophosphorylase DKFZp686D13177 EC 2.4.2.7 MGC125856 MGC125857 MGC129961 transphosphoribosidase; AMP diphosphorylase; AMP pyrophosphorylase; APRT; transphosphoribosidase
Gene Aliases: AMP; APRT; APRTD
UniProt ID: (Human) P07741
Entrez Gene ID: (Human) 353
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.