OriGene
Manufacturer Code:TA500074
Catalog # 50-167-1384
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3H3 |
Immunogen | Recombit protein expressed in E.coli corresponding to amino acids 23-188 of human APOM |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: alternative name: G3a, NG20; Apo-M; Apolipoprotein M; G3a HSPC336 NG20 apo-M; NG20-like protein; Protein G3a
Gene Aliases: apo-M; APOM; G3A; HSPC336; NG20
UniProt ID: (Human) O95445
Entrez Gene ID: (Human) 55937
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.