Abnova
Manufacturer Code:PAB7182
Catalog # 89-133-022
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human APOM. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of APOM.
For Research Use Only
Protein Aliases: alternative name: G3a, NG20; Apo-M; Apolipoprotein M; G3a; HSPC336; MGC22400; NG20; NG20-like protein; Protein G3a
Gene Aliases: apo-M; APOM; G3A; HSPC336; NG20
UniProt ID: (Human) O95445
Entrez Gene ID: (Human) 55937
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.