Proteintech
Manufacturer Code:128171AP150UL
Catalog # 50-172-8272
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | APOM Fusion Protein Ag3137 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alternative name: G3a, NG20; Apo M apolipoprotein M APOM G3a HSPC336 NG20 Protein G3a; Apo-M; Apolipoprotein M; NG20-like protein; Protein G3a; Protein Px
Gene Aliases: 1190010O19Rik; apo-M; APOM; G3A; HSPC336; NG20
UniProt ID: (Human) O95445, (Mouse) Q9Z1R3, (Rat) P14630
Entrez Gene ID: (Human) 55937, (Mouse) 55938, (Rat) 55939
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.