Proteintech
Manufacturer Code:114862AP150UL
Catalog # 50-557-076
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | APOL1 Fusion Protein Ag2016 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: APO L APOL APOL I APOL1 Apolipoprotein L Apolipoprotein L I apolipoprotein L 1 Apolipoprotein L1; Apo-L; ApoL-I; Apolipoprotein L; apolipoprotein L 1; apolipoprotein L, 1; Apolipoprotein L-I; Apolipoprotein L1
Gene Aliases: APO-L; APOL; APOL-I; APOL1; FSGS4
UniProt ID: (Human) O14791
Entrez Gene ID: (Human) 8542
Molecular Function:
apolipoprotein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.