OriGene
Manufacturer Code:CF807230
Catalog # 50-166-8169
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI10A6 |
Immunogen | Human recombit protein fragment corresponding to amino acids 120-345 of human APOH produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Activated protein C-binding protein; Anticardiolipin cofactor; APC inhibitor; Apo-H; Apolipoprotein H; apolipoprotein H (beta-2-glycoprotein I); B2G1 B2GP1 BG; B2GPI; beta(2)GPI; Beta-2-glycoprotein 1; Beta-2-glycoprotein I
Gene Aliases: APOH; B2G1; B2GP1; BG
UniProt ID: (Human) P02749
Entrez Gene ID: (Human) 350
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.