Invitrogen
Manufacturer Code:PA586687
Catalog # PIPA586687
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 264-309 of Human ApoE. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Apo-E; Apolipoprotein E; Apolipoprotein E Apo-E APOE; apolipoprotein E3
Gene Aliases: AD2; AI255918; APO-E; APOE; APOEA; LDLCQ5; LPG
UniProt ID: (Human) P02649, (Mouse) P08226, (Rat) P02650
Entrez Gene ID: (Human) 348, (Mouse) 11816, (Rat) 25728
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.