Abnova
Manufacturer Code:PAB14770
Catalog # 89-117-252
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Native purified human APOC3. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C on dry atmosphere. After reconstitution with 500&muL deionized water,store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against native APOC3.
For Research Use Only
Protein Aliases: Apo-CIII; apoC-III; APOCIII; Apolipoprotein C-III; Apolipoprotein C3; MGC150353
Gene Aliases: APOC3; APOCIII; HALP2
UniProt ID: (Human) P02656
Entrez Gene ID: (Human) 345
Molecular Function:
apolipoprotein
signaling molecule
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.