Invitrogen
Manufacturer Code:PA531132
Catalog # PIPA531132
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 34 and 263 of Human APOBEC3F |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A3F; apolipoprotein B editing enzyme catalytic polypeptide-like 3F; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3F; apolipoprotein B mRNA editing enzyme cytidine deaminase; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3F; Apolipoprotein B mRNA-editing enzyme catalytic polypeptide-like 3F; ARP8 BK150C2.4.MRNA KA6 DNA dC->dU-editing enzyme APOBEC-3F apolipoprotein B mRNA editing enzyme cytidine deaminase apolipoprotein B mRNA-editing enzyme catalytic polypeptide-like 3F induced upon T-cell activation; DNA dC->dU-editing enzyme APOBEC-3F; induced upon T-cell activation
Gene Aliases: A3F; APOBEC3F; ARP8; BK150C2.4.MRNA; KA6
UniProt ID: (Human) Q7Z2N5
Entrez Gene ID: (Human) 200316
Molecular Function: deaminase hydrolase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.