Novus Biologicals
Manufacturer Code:NBP15751320UL
Catalog # NBP15751320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to APOBEC3F(apolipoprotein B mRNA editing enzyme catalytic polypeptide-like 3F) The peptide sequence was selected from the N terminal of APOBEC3F. Peptide sequence MKPHFRNTVERMYRDTFSYNFYNRPILSRRNTVWLCYEVKT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A3F; apolipoprotein B editing enzyme catalytic polypeptide-like 3F; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3F; apolipoprotein B mRNA editing enzyme catalytic polypeptide-like 3F Apolipoprotein B mRNA-editing enzyme catalytic polypeptide-like 3F ARP8 BK150C2.4.MRNA DNA dC->dU-editing enzyme APOBEC-3F EC 3.5.4 EC 3.5.4.- induced upon T-cell activation KA6 MGC74891; apolipoprotein B mRNA editing enzyme cytidine deaminase; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3F; Apolipoprotein B mRNA-editing enzyme catalytic polypeptide-like 3F; DNA dC->dU-editing enzyme APOBEC-3F; induced upon T-cell activation
Gene Aliases: A3F; APOBEC3F; ARP8; BK150C2.4.MRNA; KA6
UniProt ID: (Human) Q7Z2N5
Entrez Gene ID: (Human) 200316
Molecular Function: deaminase hydrolase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.