Novus Biologicals
Manufacturer Code:NBP157440
Catalog # NBP157440
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to APOBEC3D (apolipoprotein B mRNA editing enzyme catalytic polypeptide-like 3D (putative)) The peptide sequence was selected from the N terminal of APOBEC3D (NP_689639). Peptide sequence: SYTWLCYEVKIKRGRSNLLWDTGVFRGP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: APOBEC3DE APOBEC3E apolipoprotein B mRNA editing enzyme catalytic polypeptide-like 3D apolipoprotein B mRNA editing enzyme catalytic polypeptide-like 3D (putative) apolipoprotein B mRNA editing enzyme catalytic polypeptide-like 3E pseudogene ARP6 DNA dC->dU-editing enzyme APOBEC-3D EC 3.5.4 EC 3.5.4.- probable DNA dC->dU-editing enzyme APOBEC-3D; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3D; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3D; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3E pseudogene; probable DNA dC->dU-editing enzyme APOBEC-3D
Gene Aliases: A3D; APOBEC3DE; APOBEC3E; ARP6
Entrez Gene ID: (Human) 140564
Molecular Function:
deaminase
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.