Invitrogen
Manufacturer Code:MA521737
Catalog # PIMA521737
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3E6 |
Immunogen | APOBEC3C (NP&lowbar055323 91 a.a. &bsim 190 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: A3C; APOBEC1-like; APOBEC3C apolipoprotein B mRNA editing enzyme catalytic polypeptide-like 3C APOBEC1L ARDC2 ARDC4 ARP5 MGC19485 PBI bK150C2.3; apolipoprotein B editing enzyme catalytic polypeptide-like 3C; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3C; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3C; DNA dC->dU-editing enzyme APOBEC-3C; Phorbolin I; probable DNA dC->dU-editing enzyme APOBEC-3C
Gene Aliases: A3C; APOBEC1L; APOBEC3C; ARDC2; ARDC4; ARP5; bK150C2.3; PBI
UniProt ID: (Human) Q9NRW3
Entrez Gene ID: (Human) 27350
Molecular Function:
deaminase
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.