Invitrogen
Manufacturer Code:PA542818
Catalog # PIPA542818
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human APOBEC3B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A3B; apolipoprotein B editing complex 3; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3B; apolipoprotein B mRNA editing enzyme catalytic polypeptide-like 3B; Apolipoprotein B mRNA editing enzyme catalytic polypeptide-like 3B APOBEC1L ARCD3 ARP4 DJ742C19.2 FLJ21201 PHRBNL bK150C 2.2; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3B; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3F; cytidine deaminase; DNA dC->dU-editing enzyme APOBEC-3B; DNA dC->dU-editing enzyme APOBEC3; phorbolin 2; phorbolin 3; Phorbolin-1-related protein; Phorbolin-2/3; probable DNA dC->dU-editing enzyme APOBEC-3B; probable DNA dC->dU-editing enzyme APOBEC3
Gene Aliases: A3B; A3Z2-Z3; A3Z2_1; A3Z3; A3Z3_1; APOBEC1L; APOBEC3; APOBEC3B; APOBEC3F; APOBEC3F1; APOBEC3F2; APOBEC3Z2; APOBEC3Z3; ARCD3; ARP4; bK150C2.2; DJ742C19.2; PHRBNL
UniProt ID: (Human) Q9UH17
Entrez Gene ID: (Human) 9582, (Pig) 100037939, (Rat) 315137
Molecular Function:
deaminase
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.