Invitrogen
Manufacturer Code:PA511430
Catalog # PIPA511430
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 299-330 amino acids from the C-terminal region of human APOBEC3B (PHO3) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A3B; apolipoprotein B editing complex 3; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3B; apolipoprotein B mRNA editing enzyme catalytic polypeptide-like 3B; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3B; Apolipoprotein B mRNA-editing complex 3; Arp3; CEM-15; CEM15; cytidine deaminase; DNA dC->dU-editing enzyme APOBEC-3; DNA dC->dU-editing enzyme APOBEC-3B; phorbolin 2; phorbolin 3; Phorbolin-1-related protein; Phorbolin-2/3; probable DNA dC->dU-editing enzyme APOBEC-3B; recovery from Friend virus 3
Gene Aliases: A3B; APOBEC1L; Apobec3; APOBEC3B; ARCD3; Arp3; ARP4; BC003314; bK150C2.2; Cem15; DJ742C19.2; PHRBNL; Rfv3
UniProt ID: (Human) Q9UH17, (Mouse) Q99J72
Entrez Gene ID: (Human) 9582, (Mouse) 80287
Molecular Function: deaminase hydrolase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.