Invitrogen
Manufacturer Code:PA578800
Catalog # PIPA578800
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human PHO1 recombit protein (Position: M1-L63). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A3A; apolipoprotein B editing complex 3; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3A; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3A; Apolipoprotein B mRNA-editing complex 3; Arp3; CEM-15; CEM15; DNA dC->dU-editing enzyme APOBEC-3A A3A Phorbolin-1 APOBEC3A; DNA dC->dU-editing enzyme APOBEC-3; DNA dC->dU-editing enzyme APOBEC-3A; phorbolin 1; Phorbolin-1; probable DNA dC->dU-editing enzyme APOBEC-3A; recovery from Friend virus 3
Gene Aliases: A3A; Apobec3; APOBEC3A; ARP3; BC003314; bK150C2.1; Cem15; PHRBN; Rfv3
UniProt ID: (Human) P31941, (Mouse) Q99J72
Entrez Gene ID: (Human) 200315, (Mouse) 80287
Molecular Function: deaminase hydrolase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.