Invitrogen
Manufacturer Code:PA525606
Catalog # PIPA525606
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 19-48 amino acids from the N-terminal region of human APOBEC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: apolipoprotein B editing complex 2; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 2; apolipoprotein B mRNA editing enzyme, catalytic polypeptide 2; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 2; ARCD1 ARP1 apolipoprotein B mRNA editing enzyme catalytic polypeptide 2 probable C->U-editing enzyme APOBEC-2; C->U-editing enzyme APOBEC-2; mRNA(cytosine(6666)) deaminase 2; probable C->U-editing enzyme APOBEC-2
Gene Aliases: APOBEC2; ARCD1; ARP1
UniProt ID: (Human) Q9Y235, (Mouse) Q9WV35
Entrez Gene ID: (Human) 10930, (Mouse) 11811
Molecular Function: deaminase hydrolase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.