Invitrogen
Manufacturer Code:PA519014
Catalog # PIPA519014
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (DVFYDPRELRKEAC) corresponding to the internal amino acids of APOBEC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: APOBEC1 apolipoprotein B mRNA editing enzyme catalytic polypeptide 1 APOBEC-1 BEDP CDAR1 HEPR apolipoprotein B mRNA editing enzyme apolipoprotein B mRNA editing enzyme complex-1; apolipoprotein B editing complex 1; Apolipoprotein B mRNA-editing enzyme 1; C->U-editing enzyme APOBEC-1; mRNA(cytosine(6666)) deaminase 1
Gene Aliases: Apobec1; Cdar1
UniProt ID: (Mouse) P51908
Entrez Gene ID: (Mouse) 11810
Molecular Function:
deaminase
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.