Proteintech
Manufacturer Code:205781AP150UL
Catalog # 50-173-2973
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Apo B 100 Apo B 48 APOB ApoB100 Apolipoprotein B Apolipoprotein B 100 FLDB; Apo B-100; Apo B-48; apolipoprotein B (including Ag(x) antigen); Apolipoprotein B-100; Apolipoprotein B-48; apolipoprotein B48
Gene Aliases: AI315052; Apo B-100; APOB; apoB-100; apoB-48; FLDB; LDLCQ4
UniProt ID: (Human) P04114, (Mouse) E9Q414
Entrez Gene ID: (Human) 338, (Mouse) 238055
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.