OriGene
Manufacturer Code:TA507140
Catalog # 50-167-6973
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A1 |
Immunogen | Full length human recombit protein of human APOA5 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Apo-AV; APOAV RAP3; Apolipoprotein A-V; apolipoprotein A-V precursor variant 3; Apolipoprotein A5; Regeneration-associated protein 3
Gene Aliases: APOA5; APOAV; RAP3; UNQ411/PRO773
UniProt ID: (Human) Q6Q788
Entrez Gene ID: (Human) 116519
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.