Abnova
Manufacturer Code:MAB10487
Catalog # 89-108-671
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1D4C11 |
Immunogen | Recombinant protein corresponding to amino acids 21-396 of human APOA4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant APOA4.
For Research Use Only
Protein Aliases: Apo-AIV; apoA-IV; Apolipoprotein A-IV; Apolipoprotein A4; MGC142154; MGC142156
Gene Aliases: APOA4
UniProt ID: (Human) P06727
Entrez Gene ID: (Human) 337
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.